Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

380687

Sigma-Aldrich

N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide

97%

Synonym(s):

N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulfonamide, N,O,O′-Tritosyldiethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2N(CH2CH2OSO2C6H4CH3)2
CAS Number:
Molecular Weight:
567.69
Beilstein:
2406580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

92-98 °C (lit.)

functional group

sulfonamide
tosylate

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCN(CCOS(=O)(=O)c2ccc(C)cc2)S(=O)(=O)c3ccc(C)cc3

InChI

1S/C25H29NO8S3/c1-20-4-10-23(11-5-20)35(27,28)26(16-18-33-36(29,30)24-12-6-21(2)7-13-24)17-19-34-37(31,32)25-14-8-22(3)9-15-25/h4-15H,16-19H2,1-3H3

InChI key

VJDZYMIEDBLSDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide may be used in the preparation of :
  • 2-(p-benzamidobenzyl)-1,4,7-tris(p-tolylsulfonyl)-1,4,7-triazacyclononane, via reaction with N,N′-bis(p-tolylsulfonyl)-1-(p-benzamidobenzyl)ethylenediamine
  • selectively protected aza-macrocycles
  • carbon-bridged metallacarboranes
  • tetrakis(phosphonomethyl)-substituted cyclododecanes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service