Skip to Content
Merck

Skip To

BAF377860000

SP Bel-Art® Spinbar® Arrester

round, diam. 2" (51 mm)

Sign Into View Organizational & Contract Pricing

Select a Size

Select a Size

Change View

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

round

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F37786-0000

diam.

2" (51 mm)

General description

The Magnetic Stirring Bar Arrester holds your stirring bar in place when pouring from a beaker or flask by placing the arrester against the bottom of the container.

Features and Benefits

  • 9.5mm (⅜") thick polyethylene
  • 51mm (2") diameter with a radial magnet
  • Other sizes can be made to order

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service