About This Item
UNSPSC Code:
41121815
eCl@ss:
32010310
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
316L stainless steel lock ring (ASME SA-479)
316L stainless steel welding flange (ASME SA-479)
stainless steel bolt (A193 Gr B8M/A194 Gr B8M)
stainless steel nut (A193 Gr B8M/A194 Gr B8M)
Quality Level
product line
NovAseptic®
parameter
pressure range at -1--7 bar
temp. range at -80-200 °C
weight
1.3 kg (2.87 lb)
Application
The applied gasket (not included) may have different design temperature and/or pressure limits than the NovAseptic® Connector. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
The NOVASEPTIC® Connector does comply for USA with the relevant requirements in ASME. The equipment has been calculated and evaluated of stress levels due to internal pressure in accordance with ASME VIII, Division 2, "Mandatory design based on stress anal