Skip to Content
Merck

Skip To

NIST277

Tungsten concentrate

NIST SRM 277

Synonym(s):

Wolframite

Sign Into View Organizational & Contract Pricing

Select a Size

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

packaging

pkg of 1 bottle x 100 g

manufacturer/tradename

NIST®

format

neat

General description

This Standard Reference Material (SRM) is a tungsten concentrate primarily derived from wolframite ores through commercial refining processes and is intended for use in the validation of chemical and instrumental methods of analysis. SRM 277_cert SRM 277 _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service