Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C25H25F3N4O6
CAS Number:
Molecular Weight:
534.48
UNSPSC Code:
51101500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

dry media

color

white to off-white

antibiotic activity spectrum

mycobacteria

Mode of action

cell wall synthesis | inhibits

shipped in

ambient

storage temp.

−20°C

SMILES string

C[C@@]1(CN2C=C(N=C2O1)[N+](=O)[O-])COC3=CC=C(C=C3)N4CCC(CC4)OC5=CC=C(C=C5)OC(F)(F)F

InChI

InChI=1S/C25H25F3N4O6/c1-24(15-31-14-22(32(33)34)29-23(31)38-24)16-35-18-4-2-17(3-5-18)30-12-10-20(11-13-30)36-19-6-8-21(9-7-19)37-25(26,27)28/h2-9,14,20H,10-13,15-16H2,1H3/t24-/m1/s1

InChI key

XDAOLTSRNUSPPH-XMMPIXPASA-N

General description

Delamanid is an anti-tuberculosis agent derived from the nitro-dihydro-imidazooxazole class of compounds that inhibits mycolic acid synthesis of bacterial cell wall. A limitation of dasabuvir is the lower potential for cross-genotypic activity as the binding sites are poorly preserved across HCV genotypes.

Biochem/physiol Actions

Delamanid is an antimycobacterial agent that inhibits mycolic acid synthesis, an essential component of the cell wall of Mycobacterium tuberculosis. Inhibition of mycolic acid synthesis leads to the disruption of cell wall structure and integrity, and ultimately, bacterial cell death.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service