Skip to Content
Merck

Skip To

SML3520

Talaporfin sodium

new

≥95% (HPLC)

Synonym(s):

Aspartyl chlorin tetrasodium, LS11 tetrasodium, ME 2906 tetrasodium, ME2906 tetrasodium, Mono-L-aspartyl chlorin tetrasodium, Mono-L-aspartyl chlorin e6 tetrasodium, NPe6 tetrasodium, Talaporfin tetrasodium, N-[[(7S,8S)-3-Carboxy-7-(2-carboxyethyl)-13-ethenyl-18-ethyl-7,8-dihydro-2,8,12,17-tetramethyl-21H,23H-porphin-5-yl]acetyl]-L-aspartic acid, tetrasodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H37N5Na4O9
CAS Number:
Molecular Weight:
799.69
MDL number:
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

, Dark green to black

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(C[C@H](NC(C/C1=C2[C@@H](CCC(O[Na])=O)[C@H](C)C(/C=C3N/C(C(C=C)=C\3C)=C\C4=N/C(C(CC)=C4C)=C\C5=C(C(C(O[Na])=O)=C1N5)C)=N\2)=O)C(O[Na])=O)O[Na]

Biochem/physiol Actions

Cancer tissue-targeting photosensitizer that induces irradiation-dependent cancer killing and in vivo photodynamic therapy (PDT) efficacy.

Talaporfin is a chlorin based cancer tissue-targeting photosensitizer that, upon irradiation, generates reactive oxygen species (ROS) that cause DNA damage and apoptosis in cancer cells. Talaporfin induces irradiation-dependent cancer killing in cultures (24 h 10-100 μg/mL Talaporfin sodium followed by washing and 664 nm laser irradation at 15 mW/cm2 and 10 J/cm2; ESCC) and displays photodynamic therapy (PDT) efficacy in human cancer exnograft models in mice in vivo, including EMT-6 sarcoma (100% cure with 664 nm laser at 100 J/cm2 and 100 mw/cm2 24h after 8 mg/kg talaporfin i.p.) and ESCC (664 nm, 100 J/cm2 2h after 10 mg/kg talaporfin sodium via tail vein i.v.).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service