Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C22H20F5N3O3
CAS Number:
Molecular Weight:
469.40
UNSPSC Code:
12352200
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Orally active, potent and selective gamma-secretase inhibitor with anti-cancer efficacy in vitro and in vivo.
RO4929097 is an orally active, potent and selective gamma-secretase (γ-secretase) inhibitor (IC50 = 4 nM) that blocks Aβ1-40 production (IC50 = 14 nM in APP-overexpressing HEK293) and suppresses Notch signaling (IC50 = 5 nM by reporter assays) with >100-fold selectivity over 75 other enzymes, receptors and ion channels. RO4929097 reduces the size of MDA-MB-468 soft agar colonies in cultures (100-250 nM) and effectively suppresses A549 NSCLC xenografts-derived tumor growth in mice in vivo (3-60 mg/kg bid or qd p.o.).
RO4929097 is an orally active, potent and selective gamma-secretase (γ-secretase) inhibitor (IC50 = 4 nM) that blocks Aβ1-40 production (IC50 = 14 nM in APP-overexpressing HEK293) and suppresses Notch signaling (IC50 = 5 nM by reporter assays) with >100-fold selectivity over 75 other enzymes, receptors and ion channels. RO4929097 reduces the size of MDA-MB-468 soft agar colonies in cultures (100-250 nM) and effectively suppresses A549 NSCLC xenografts-derived tumor growth in mice in vivo (3-60 mg/kg bid or qd p.o.).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service