Product Name
Ethylaluminum dichloride solution 25 wt. % in toluene
Product Number
192759
Product Brand
ALDRICH
CAS Number
563-43-9
Molecular Weight
126.95
Test | Specification |
---|---|
Appearance (Color) | Colorless to Yellow |
Appearance (Form) | Liquid |
Purity Titration by EDTA (M) | 23.0 - 27.0 M |
Complexometric |