Product Name
Trichloroacetyl isocyanate 96%
Product Number
217328
Product Brand
ALDRICH
CAS Number
3019-71-4
Molecular Weight
188.40
Test | Specification |
---|---|
APPEARANCE | COLORLESS LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 95.5% (MINIMUM) |
APPROVED FEBRUARY 14, 2006 RJM |