Product Name
Tetraphenylphosphonium chloride 98%
Product Number
218790
Product Brand
ALDRICH
CAS Number
2001-45-8
Molecular Weight
374.84
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE TO BEIGE POWDER OR |
GRANULAR POWDER AND/OR CHUNKS | |
IDENTITY | CONFORMS TO STRUCTURE BY IR OR HNMR SPECTRA. |
TITRATION | 97.5% - 102.5% (WITH SILVER NITRATE) |
REVISED APRIL 4, 2008 BTR |