Product Name
Carvacrol 98%
Product Number
282197
Product Brand
ALDRICH
CAS Number
499-75-2
Molecular Weight
150.22
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO ORANGE LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD. |
GAS LIQUID | 97.5% (MINIMUM) |
APPROVED FEBRUARY 27, 1996 JSB |