Product Name
Methylmagnesium bromide solution 1.4 M in THF: toluene (1:3)
Product Number
282235
Product Brand
ALDRICH
CAS Number
75-16-1
Molecular Weight
119.24
Test | Specification |
---|---|
APPEARANCE | LIGHT TAN TO LIGHT YELLOW, ORANGE, BROWN OR |
BLACK LIQUID | |
TITRATION | 1.33M - 1.54M (WITH 2-BUTANOL) |
APPROVED JULY 16, 1998 JSB |