Product Name
N-Methoxy-N-methylbenzamide 98%
Product Number
345415
Product Brand
ALDRICH
CAS Number
6919-61-5
Molecular Weight
165.19
Test | Specification |
---|---|
Appearance (Color) | Colorless to Yellow |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥97.5 % |