Product Name
Serinol 98%
Product Number
357898
Product Brand
ALDRICH
CAS Number
534-03-2
Molecular Weight
91.11
Test | Specification |
---|---|
Appearance (Color) | Conforms to Requirements |
White to Off-White or Light Yellow | |
Appearance (Form) | Conforms to Requirement |
Crystals or Crystalline Powder | |
Infrared spectrum | Conforms to Structure |
Purity (Titration by HCL04) | 97.5 - 102.5 % |