Product Name
1-Iodo-1H,1H,2H,2H-perfluorodecane 96%
Product Number
370525
Product Brand
ALDRICH
CAS Number
2043-53-0
Molecular Weight
574.02
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN SOLID, POWDER AND/OR CHUNKS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 95.5-104.5% (WITH AGNO3 AFTER O2 COMBUSTION) |
GAS LIQUID | 95.5% (MINIMUM) |
APPROVED JANUARY 30, 2008 RJM |