Product Name
(DHQ)2PHAL ≥95%
Product Number
392723
Product Brand
ALDRICH
CAS Number
140924-50-1
Molecular Weight
778.98
Test | Specification |
---|---|
Appearance (Color) | White |
Appearance (Form) | Crystal to Powder |
Infrared spectrum | Conforms to Structure |
Proton NMR spectra | Conforms to Structure |
Carbon | 70.3 - 77.7 % |
Nitrogen | 10.2 - 11.3 % |
Optical Rotation | 316 - 356 ° |
c = 1.2%; Methanol | |
Purity (HPLC) | ≥95.0 % |