Product Name
2-(4-Benzoyl-3-hydroxyphenoxy)ethyl acrylate 98%
Product Number
413216
Product Brand
ALDRICH
CAS Number
16432-81-8
Molecular Weight
312.32
Test | Specification |
---|---|
APPEARANCE | WHITE TO LIGHT YELLOW POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 97.5% (MINIMUM) |
REVISED APRIL 28, 2008 BTR |