Product Name
N-Benzylethylenediamine 97%
Product Number
462292
Product Brand
ALDRICH
CAS Number
4152-09-4
Molecular Weight
150.22
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5%-103.5% (WITH HCLO4) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED OCTOBER 17, 2007 RJM |