Product Name
Cyclopropylacetonitrile 97%
Product Number
516112
Product Brand
ALDRICH
CAS Number
6542-60-5
Molecular Weight
81.12
Test | Specification |
---|---|
APPEARANCE | COLORLESS LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED NOVEMBER 16, 2006 RJM |