Product Name
Poly(L-lactide-co-caprolactone-co-glycolide) L-lactide 70 %, average Mn ~50,000 by GPC
Product Number
568562
Product Brand
ALDRICH
CAS Number
134490-19-0
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | WHITE SOLID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GEL PERMEATION | MW= CA 80,000 |
CHROMATOGRAPHY | MN= CA 50,000 |
PI= CA 2.0 | |
PROTON NMR SPECTRUM | APPROXIMATE COMPOSITION (MOLE %) WILL BE |
REPORTED EACH LOT, TYPICALLY | |
LL:CL:G = 70:20:10 | |
REVISED AUGUST 30, 2007 JSB |