Product Name
α-Cyclopentylmandelic acid 97%
Product Number
706396
Product Brand
ALDRICH
CAS Number
427-49-6
Molecular Weight
220.26
Test | Specification |
---|---|
Appearance (Color) | White to Tan |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Purity (Titration by NaOH) | 96.5 - 103.5 % |
GC (area %) | ≥96.5 % |