Product Name
Butyltrimethylammonium bis(trifluoromethylsulfonyl)imide 99%
Product Number
713007
Product Brand
ALDRICH
CAS Number
258273-75-5
Molecular Weight
396.37
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow |
APPEARANCE (FORM) | Liquid |
TITRATION (ION EXCHANGE) | 98.5 - 101.5 % |
REFRACTIVE INDEX N20/D | 1.400 - 1.420 |
WATER | ≤ 0.2 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
CARBON 13 NMR SPECTRUM | CONFORMS TO STRUCTURE |
ION CHROMATOGRAPHY (BROMIDE) | ≤ 25 mg/kg |
ION CHROMATOGRAPHY (CHLORIDE) | ≤ 100 mg/kg |
ION CHROMATOGRAPHY (FLUORIDE) | ≤ 25 mg/kg |
ION CHROMATOGRAPHY (NITRATE) | ≤ 25 mg/kg |
ION CHROMATOGRAPHY (PHOSPHATE) | ≤ 10 mg/kg |
ION CHROMATOGRAPHY (SULFATE) | ≤ 25 mg/kg |
RECOMMENDED RETEST PERIOD | 48 MONTHS |