Product Name
N,N′-Dimethylthiourea 99%
Product Number
D188700
Product Brand
ALDRICH
CAS Number
534-13-4
Molecular Weight
104.17
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER, CRYSTALS, |
CRYSTALLINE POWDER OR SOLID AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 98.5% - 101.5% (WITH BA(CLO4)2, AFTER OXYGEN |
COMBUSTION) | |
AND/OR | |
ELEMENTAL ANALYSIS | CARBON 34.0%-35.1% |
NITROGEN 26.4%-27.3% | |
THIN-LAYER | CONSISTENT WITH 99% PURITY |
SOLUBILITY | 25MG/ML (2.5%) METHANOL; CLEAR, COLORLESS |
SOLUTION | |
REVISED FEBRUARY 8, 2005 RJM |