Product Name
trans-3-Indoleacrylic acid 98%
Product Number
I3807
Product Brand
ALDRICH
CAS Number
29953-71-7
Molecular Weight
187.19
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 98.5% - 101.5% (WITH NAOH) |
THIN-LAYER | CONSISTENT WITH 99% IMPURITY |
OR | |
HIGH PRESSURE LIQUID | 98.5% (MINIMUM) |
SOLUBILITY | 50MG/ML, ACETONE; CLEAR TO HAZY, YELLOW TO |
YELLOW-ORANGE SOLUTION | |
REVISED SEPTEMBER 12, 2006 RJM |