Product Name
1-Octadecanol 95%
Product Number
O709
Product Brand
ALDRICH
CAS Number
112-92-5
Molecular Weight
270.49
Test | Specification |
---|---|
APPEARANCE | WHITE FLAKES |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 113C OF EDITION I, | |
VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
GAS LIQUID | 94.0% (MINIMUM) |
APPROVED DECEMBER 6, 1995 JSB |