Product Name
Propionyl chloride 98%
Product Number
P51559
Product Brand
ALDRICH
CAS Number
79-03-8
Molecular Weight
92.52
Test | Specification |
---|---|
APPEARANCE | COLORLESS LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5%-102.5%(WITH AGNO3 AFTER O2 COMBUSTION) |
GAS LIQUID | 97.5% (MINIMUM) |
REVISED FEBRUARY 6, 2002 JSB |