Product Name
Conjugated (9Z,11E)-Linoleic acid analytical standard
Product Number
16413
Product Brand
SIAL
CAS Number
2540-56-9
Molecular Weight
280.45
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow and Colorless to Light Green-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (HPLC AREA %) | ≥ 96.0 % |
REFRACTIVE INDEX N20/D | 1.478 - 1.483 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 18 MONTHS |