Product Name
Terbumeton PESTANAL®, analytical standard
Product Number
31527
Product Brand
SIAL
CAS Number
33693-04-8
Molecular Weight
225.29
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Yellow to Faint Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
MELTING POINT | 118 - 124 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |