Product Name
Parbendazole VETRANAL®, analytical standard
Product Number
32438
Product Brand
SIAL
CAS Number
14255-87-9
Molecular Weight
247.29
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
WATER | ≤ 3.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 48 MONTHS |