Product Name
Robenidine hydrochloride VETRANAL®, analytical standard
Product Number
33979
Product Brand
SIAL
CAS Number
25875-50-7
Molecular Weight
370.66
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
WATER | ≤ 5.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |