Product Name
PCB No 15 analytical standard
Product Number
35597
Product Brand
SIAL
CAS Number
2050-68-2
Molecular Weight
223.10
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Orange |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 98.0 % |
IDENTIFICATION (GC-MS) | CORRESPONDS TO REQUIREMENTS |
MELTING POINT | 145 - 150 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |