Product Name
2-Methylnaphthalene analytical standard
Product Number
45796
Product Brand
SIAL
CAS Number
91-57-6
Molecular Weight
142.20
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Yellow and Faint Beige to Beige and Faint Brown to Light Brown and Yellow-Brown |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 98.0 % |
MELTING POINT | 31 - 37 C |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |