Product Name
1,2,3-Trimethylbenzene analytical standard
Product Number
45935
Product Brand
SIAL
CAS Number
526-73-8
Molecular Weight
120.19
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 90.0 % |
REFRACTIVE INDEX N20/D | 1.5121 - 1.5161 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |