Product Name
Esfenvalerate PESTANAL®, analytical standard
Product Number
46277
Product Brand
SIAL
CAS Number
66230-04-4
Molecular Weight
419.90
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 98.0 % |
SPECIFIC ROTATION (20/D) | -12.4 ± 2.0 Degrees |
CONCENTRATION | C=1 in CHLOROFORM |
MELTING POINT | 58 - 64 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |