Product Name
o-Hydroxyhippuric acid analytical standard
Product Number
49861
Product Brand
SIAL
CAS Number
487-54-7
Molecular Weight
195.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Yellow |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 97.0 % |
CARBON CONTENT | 55.39% (Theory) |
HYDROGEN CONTENT | 4.65% (Theory) |
NITROGEN CONTENT | 7.18% (Theory) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
REMARKS | ≤ 1.0% WATER (calculated from elemental analysis) |
RECOMMENDED RETEST PERIOD | 36 MONTHS |