Product Name
Triethylamine hydrochloride suitable for HPLC, LiChropur™, 99.0-101.0% (AT)
Product Number
63961
Product Brand
SIAL
CAS Number
554-68-7
Molecular Weight
137.65
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
ARGENTOMETR. TITRATION | 99.0 - 101.0 % |
SUITABILITY TEST (HPLC) | CORRESPONDS TO STANDARD |
MELTING POINT (DEC.) | 256 - 259 C |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 1g in 10ml water |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
UV Absorbance | UV-Absorption at 220 nm: ≤ 0.05 |
UV Absorbance | UV-Absorption at 230 nm: ≤ 0.04 |
UV Absorbance | UV-Absorption at 250 nm: ≤ 0.03 |
UV Absorbance | UV-Absorption at 260 nm: ≤ 0.02 |
UV Absorbance | UV-Absorption at 500 nm: ≤ 0.02 |
CYCLIC VOLTAMMOGRAM | CORRESPONDS TO REQUIREMENTS |
RESIDUE (FILTER TEST) | NO RESIDUE |
FILTERTEST (METHOD) | 3g in 30ml water |