Product Name
Potassium phosphate dibasic trihydrate puriss. p.a., ≥99.0% (T)
Product Number
60349
Product Brand
SIGALD
CAS Number
16788-57-1
Molecular Weight
228.22
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Crystals |
TITRATION | 99.0 - 101.0 % |
TITRATION (METHOD) | BACK TITRATION |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 1M IN WATER |
PH-TEST | 9.1 - 9.4 |
ARSEN | ≤ 0.00001 % |
CALCIUM | ≤ 0.001 % |
CADMIUM | ≤ 0.0005 % |
COBALT | ≤ 0.0005 % |
CHROMIUM | ≤ 0.0005 % |
COPPER | ≤ 0.0005 % |
IRON | ≤ 0.0005 % |
MAGNESIUM | ≤ 0.0005 % |
MANGANESE | ≤ 0.0005 % |
SODIUM | ≤ 0.05 % |
NICKEL | ≤ 0.0005 % |
LEAD | ≤ 0.0005 % |
ZINC | ≤ 0.0005 % |
CHLORIDE (CL) | ≤ 20 mg/kg |
TOTAL NITROGEN | ≤ 10 mg/kg |
SULFATE (SO4) | ≤ 50 mg/kg |
RECOMMENDED RETEST PERIOD | 60 Months |