Product Name
L-Arginine BioUltra, ≥99.5% (NT)
Product Number
11009
Product Brand
SIGMA
CAS Number
74-79-3
Molecular Weight
174.20
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION (NT) HCLO4 0.1M | 99.5 - 100.5 % |
PURITY (TLC AREA %) | ≥ 99.7 % |
SPECIFIC ROTATION (20/D) | 27.0 ± 0.5 Degrees |
CONCENTRATION | C=5 IN HYDROCHLORIC ACID 5M |
LOSS ON DRYING | ≤ 0.1 % |
SULFATED ASH | ≤ 0.05 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
ALUMINIUM (ICP) | ≤ 5 mg/kg |
BARIUM (ICP) | ≤ 5 mg/kg |
BISMUTH (ICP) | ≤ 5 mg/kg |
CALCIUM (ICP) | ≤ 10 mg/kg |
CADMIUM (ICP) | ≤ 5 mg/kg |
COBALT (ICP) | ≤ 5 mg/kg |
CHROMIUM (ICP) | ≤ 5 mg/kg |
COPPER (ICP) | ≤ 5 mg/kg |
IRON (ICP) | ≤ 5 mg/kg |
POTASSIUM (ICP) | ≤ 50 mg/kg |
LITHIUM (ICP) | ≤ 5 mg/kg |
MAGNESIUM (ICP) | ≤ 5 mg/kg |
MANGANESE (ICP) | ≤ 5 mg/kg |
MOLYBDENUM (ICP) | ≤ 5 mg/kg |
SODIUM (ICP) | ≤ 50 mg/kg |
NICKEL (ICP) | ≤ 5 mg/kg |
LEAD (ICP) | ≤ 5 mg/kg |
STRONTIUM (ICP) | ≤ 5 mg/kg |
ZINC (ICP) | ≤ 5 mg/kg |
ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
TOTAL SULFUR AS SO4 (ICP) | ≤ 100 mg/kg |
CHLORIDE (CL) | ≤ 100 mg/kg |
SOLUBILITY (METHOD) | 2.613 G IN 30 ML H2O (0.5 M) |
APPEARANCE (SOLUTION) | CLEAR COLORLESS |
PH (SOLUTION) | 10.5 - 12.0 |
RESIDUE (FILTER TEST) | NO RESIDUE |
UV - ABS. AT 260 NM | ≤ 0.2 |
UV - ABS. AT 280 NM | ≤ 0.1 |
RECOMMENDED RETEST PERIOD | 72 MONTHS |