Product Name
Brij® L23 main component: tricosaethylene glycol dodecyl ether
Product Number
16005
Product Brand
SIGMA
CAS Number
9002-92-0
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Pellets |
MELTING POINT | 39 - 43 C |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear to Slightly Hazy |
SOLUBILITY (METHOD) | 0.4 G IN 10ML WATER, HOT |
CARBON CONTENT | FOR INFORMATION |
HYDROGEN CONTENT | FOR INFORMATION |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
CALCIUM (ICP) | ≤ 10 mg/kg |
CADMIUM (ICP) | ≤ 5 mg/kg |
COBALT (ICP) | ≤ 5 mg/kg |
CHROMIUM (ICP) | ≤ 5 mg/kg |
COPPER (ICP) | ≤ 5 mg/kg |
IRON (ICP) | ≤ 5 mg/kg |
POTASSIUM (ICP) | ≤ 500 mg/kg |
MAGNESIUM (ICP) | ≤ 5 mg/kg |
MANGANESE (ICP) | ≤ 5 mg/kg |
SODIUM (ICP) | ≤ 500 mg/kg |
NICKEL (ICP) | ≤ 5 mg/kg |
LEAD (ICP) | ≤ 5 mg/kg |
ZINC (ICP) | ≤ 5 mg/kg |