Product Name
Malachite Green oxalate salt suitable for microscopy, crystalline, S. No.: 754
Product Number
32745
Product Brand
SIGMA
CAS Number
2437-29-8
Molecular Weight
463.50
Test | Specification |
---|---|
APPEARANCE (COLOR) | Green to Very Dark Green |
APPEARANCE (FORM) | Powder or Crystals |
THIN LAYER CHROMATOGR. | CORRESPONDS TO STANDARD |
SOLUBILITY (COLOR) | 0.01G IN 10ML METHANOL |
SOLUBILITY (METHOD) | 0.01G IN 10ML METHANOL |
LOSS ON DRYING | ≤ 7.0 % |
WAVELENGTH (1) (UV) | 609 - 619 nm |
EXTINCTION (1%/1CM) (1) (UV) | ≥ 1650 |
WAVELENGTH (2) (UV) | 420 - 430 nm |
EXTINCTION (1%/1CM) (2) (UV) | ≥ 300 |
SOLVENT (UV) | Water |