Product Name
2-Octyl-4-isothiazolin-3-one PESTANAL®, analytical standard
Product Number
46078
Product Brand
SIGMA
CAS Number
26530-20-1
Molecular Weight
213.34
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Green to Light Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (HPLC AREA %) | ≥ 98.0 % |
REFRACTIVE INDEX N20/D | 1.5198 - 1.5238 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |