Product Name
Pectinase from Aspergillus ≥0.3 U/mg
Product Number
89216
Product Brand
SIGMA
CAS Number
9032-75-1
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Off White to Light Yellow to Light Brown and Faint Grey to Light Grey |
APPEARANCE (FORM) | Powder |
SOLUBILITY (COLOR) | Report Result |
SOLUBILITY (TURBIDITY) | Report Result |
SOLUBILITY (METHOD) | 5 MG/ML IN ENZYME DILUTION BUFFER |
ENZYMATIC ACTIVITY | ≥ 0.3 U/mg |
SPECIFIC ACTIVITY | 1 U corresponds to the amount of enzyme which liberates 1 umol galacturonic acid from polygalacturonic acid per minute at pH 4.0 and 50 deg C. |
RECOMMENDED RETEST PERIOD | 48 MONTHS |