Product Name
Boc-Glu-OBzl
Product Number
B9003
Product Brand
SIGMA
CAS Number
30924-93-7
Molecular Weight
337.37
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
SOLUBILITY | CLEAR TO SLIGHTLY HAZY COLORLESS TO LIGHT YELLOW SOLUTION AT 50 MG/ML IN ETHANOL |
IR SPECTRUM | CONSISTENT WITH STRUCTURE |
CARBON | 59.0 TO 62.0% |
NITROGEN | 3.8 TO 4.5% |
SPECIFIC ROTATION | -29.0 +/- 1 DEG (C = 1 IN METHANOL AT 24 DEG C) * |
PURITY BY THIN LAYER CHROMATOGRAPHY | >OR = 98% |
DOCUMENT # | B9003/10/6/08/1 |