Product Name
Nutrient Mixture F-10 Ham With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Product Number
N6908
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | CLEAR SOLUTION WITH NO PARTICULATES OR PRECIPITATES |
PH TEST AT RT | 7.0 - 7.6 |
OSMOLALITY TEST | 271 - 299 MOSM/KG |
STERILITY BY MF | STERILE |
ENDOTOXIN ASSAY | NMT 1.0 EU/ML |
GLUCOSE | 0.99 - 1.21 G/L |
AMINO ACIDS BY HPLC | CONSISTENT WITH FORMULA |
KEY ELEMENTS BY ICP | CONSISTENT WITH FORMULA |
CELL CULTURE TEST | PASS |
EXPIRATION DATE | 12 MONTHS |
N6908:05/00-1 |