Product Name
L-Pyroglutamic acid 2-naphthylamide ≥99% (TLC)
Product Number
P5891
Product Brand
SIGMA
CAS Number
22155-91-5
Molecular Weight
254.28
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS TO VERY SLIGHTLY HAZY COLORLESS SOLUTION AT 50MG/ML IN METHANOL |
IR SPECTRUM | CONSISTENT WITH STRUCTURE |
ELEMENTAL ANALYSIS | 69.4 TO 72.3% CARBON |
10.7 TO 11.3% NITROGEN | |
PURITY BY THIN LAYER CHROMATOGRAPHY | GREATER THAN 99% |
RECOMMENDED RETEST | 3 YEARS |
DOCUMENT # P5891/04/19/00/2 |