Product Name
Superose® 12 Prep Grade
Product Number
S9514
Product Brand
SIGMA
CAS Number
97599-42-3
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | WHITE SUSPENSION |
PARTICLE SIZE DISTRIBUTION | > OR = 80% BETWEEN 20 AND 40 MICRONS |
GEL CONTENT | 1 ML PACKED GEL PER 1.0 TO 1.4 ML SUSPENSION |
SUPEROSE IS A REGISTERED TRADEMARK OF GE HEALTHCARE | |
DOCUMENT # | S9514/02/16/09/1 |