Product Name
Anti-Perilipin, C-Terminal antibody produced in goat affinity isolated antibody, buffered aqueous solution
Product Number
SAB2500775
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | 100µg/200µL |
ELISA (Peptide) | Antibody detection limit dilution 1:32000. |
Western Blotting (Cell lysate) | Approx. 55-60kDa band observed in Human Adipose lysates. Recommended concentration: 0.01-0.03 µg/ml. |
Immunohistochemistry - Paraffin Embedded Tissue | Not Verified |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |