Product Name
Anti-GPR37 (AB2) antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
SAB2900130
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Liquid |
Concentration | 1 mg/ml |
ELISA | Not Verified |
Western Blotting | Not Verified |
Immunohistochemistry | Antibody concentration: 20 µg/ml |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |