Product Name
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF™ 488A antibody produced in goat ~2 mg/mL, affinity isolated antibody
Product Number
SAB4600045
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Absorption Maxium | 490 ± 5nm |
Degree of labeling | 6-10 |
Antibody reactivity | Rabbit IgG² |
Purity | Negligible or no free dye detected |