Product Name
Monoclonal Anti-ROR1 antibody produced in mouse clone 2H6, ascites fluid
Product Number
SAB5300209
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | light yellow liquid |
Concentration | Not Available |
ELISA Antibody Concentration | Antibody concentration: 1/10000, using recombinant protein. |
Western Blotting (Recombinant Protein) | Antibody concentration: 1/500 - 1/2000. |
Western Blotting (Cell Lysate) | Antibody concentration: 1/500 - 1/2000. |
Immunohistochemistry | Not Determined |
Immunofluorescence | Antibody concentration: 1/200 - 1/1000. |
Immunoprecipitation | Not Determined |
Flow Cytometry | Not Determined |
Chromatin Immunoprecipitation | Not Determined |